Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meridian Bioscience™ Para-Pak SpinCon™ Concentration System
Requires only one spin for recovery of protozoan cysts, trophs and helminth ova and larvae
Supplier: Meridian Bioscience™ 972200
Description
- Ethyl acetate-free
- Based on filtration requiring no chemicals
- Contains Spincon devices, funnels, caps 15mL tubes and surfactant
- Stool specimen
- Shelf life of 36 months
Certifications
CE marked
Specifications
Tube | |
One-spin Specimen Concentration System | |
Spincon devices, 15mL tubes, funnels, caps and surfactant |
Stool | |
Concentrates eggs, larvae and protozoa from preserved stools | |
200 Case |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction