Learn More
Meridian Bioscience™ Para-Pak™ EcoFix™ Stool Transportation and Fixative Systems
SDP
Formalin-free fixative
Supplier:  Meridian Bioscience™ 981312
 View more versions of this product
Catalog No. 14-910-127
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
Includes: A spork for specimen manipulation, a squeezable vial for rapid specimen filtration and a modified cap that adapts to 15- or 50mL conical processing tubes
Description
Description
  • Designed for collecting, transporting, filtering, preserving and examining stool specimens for intestinal parasites
  • Works with trichrome and iron hematoxylin stain procedures
  • Preservative eliminates the need for processing in a hood, reduces disposal cost and minimizes the adverse effects from delay in specimen transport
  • Ideal for the recovery of eggs and cysts in concentration procedures
  • Both concentration and permanent stain may be performed on the EcoFix-preserved specimen
  • For each of the systems, there is also a corresponding clean system kit
EcoFix Fixative System A single vial containing EcoFix zinc-based preservative designed for collecting, transporting, filtering and preserving fecal parasites EcoFix Fixative/Clean Vial System A two-vial system including an EcoFix vial for processing preserved fecal specimens and a leakproof, 30mL clean vial for transporting specimens that do not need a preservative transport media
EcoFix ULTRA System Combines the concentration benefits of the Para-Pak ULTRA with the formalin-free EcoFix fixative. In the ULTRA format, a filter is incorporated into the vial cap.
EcoFix ULTRA/Clean Vial System Two-vial system includes both an EcoFix ULTRA vial (described above) for processing preserved fecal specimens and a clean vial for processing fecal specimens that have not been preserved.
Specifications
Specifications
Stool Transport System
Specimen manipulation spork, squeezable vial, modified cap (15 or 50mL conical processing tubes)
1
20°C to 30°C
CE
Ultra EcoFix Fixative
120/Cs.
Stool
Safety and Handling
Safety and Handling
ShelfLife : 2 Years