Learn More
Meridian Bioscience™ GullSORB™ IgG Inactivation Reagent
Diluted reagent for the inactivation of IgG antibodies in human serum
Supplier: Meridian Bioscience™ XX715
Alert:
Contains sodium azide, which may form explosive azide salts if allowed to build up in lead or copper plumbing. When disposing of the reagent in sinks, be sure to flush with a large colume of water.
Sodium azide is very toxic in case of ingestion. In contact with acid it emits a very toxic gas. In case of contact with the skin wash immediately and abundantly with water.
Description
GullSORB™ Reagent for the inactivation of IgG antibodies in human serum
- Reagent provides a means of eliminating both types of intereference caused by immune IgG thorugh an immunoprecipitation with antihuman IgG antibodies
- Patient sera treated with GullSORB Reagent can be tested for IgM with confidence in the accuracy of the assay
- Supplied as a ready-to-use liquid
- Should be stored at 2-8°C and should not be used beyond the expiration date on the label (24 months from the time of manufacture)
Specifications
Liquid | |
Sodium chloride 0.38%, sodium azide 0.10% | |
5mL | |
2 min. |
Inactivation of IgG antibodies in human serum | |
2° to 8°C | |
IgG Inactivation Reagent |
Safety and Handling
ShelfLife : 36 Months
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.