Learn More
Molecular Toxicology mEndo Les Agar, 50 mm Plate, 8 mL/plate, 10 plates/sleeve, Store in Refrigerator
Supplier: Molecular Toxicology 2140S204
m Endo Agar, LES is used for enumerating coliforms in water by membrane filtration technique. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.