Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Men and Women of Science VHS Video
Behind most major scientific discoveries is a fascinating story.
Specifications
Format | VHS |
---|---|
Includes | Teacher's Guide |
Grade | 7 to 12 |
Includes: Teacher's guide with background information on the scientist, “what to watch for” questions, discussion topics, and suggestions for follow-up activities. Also includes black line reproducible activity sheets stored in a sturdy vinyl binder.
Catalog Number | Mfr. No. | Name | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Name | Price | Quantity & Availability | |||||
S88091
|
*IMPERIAL INTERNATIONAL
IIMW3000 |
Biology Set of 5 VHS | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
S88096
|
*IMPERIAL INTERNATIONAL
II MW 16 |
Nutrition VHS | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
S88099
|
*IMPERIAL INTERNATIONAL
II MW 10 |
Chemistry of Burning VHS | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
S88102
|
*IMPERIAL INTERNATIONAL
II MW 1 |
Sorting VHS | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Students in elementary grades and above are treated to a fascinating program that goes beyond coverage of a particular major scientific discovery to a look at the life and work of the scientist behind it
- These 18 videos offer a unique forum for learning
- Grades 1-6, remedial for Grades 7-12
Order Info
Sold separately or in topic sets.
Specifications
VHS | |
Teacher's Guide | |
7 to 12 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title