Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Cut Pro™ 92723PU HyperMax™ Gloves, Black Polyurethane Coating

Catalog Number p-7138432
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Salt and pepper colored HyperMax™ shell features black polyurethane coating on palm and fingertips for cut, tear and abrasion resistance.

  • Salt and pepper colored 13-gauge HyperMax™ shell
  • Black polyurethane coated palm and fingertips
  • Seamless shell is comfortable, cool and easy to wear
ANSI Abrasion Rating ANSI 4
Certifications/Compliance ANSI Cut 2
Cuff Material HPPE
Coating Material Polyurethane
Color Black, Multicolored
Cut Resistance Rating ANSI A2
Material HPPE
Product Type Cut-resistant Gloves
Size X-Small
For Use With (Application) Assembly Line Production, Contruction, Material Handling
Glove Cuff Color Multi-colored
Glove Cuff Style Knit
Hem Color Multi-colored
Model 92723PUXS
Series Cut Pro™ 92723PU
Yarn Gauge 13
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit