Learn More
Stanford Research Systems Inc MELTING POINT APPARATUS (WITH
Supplier: Stanford Research Systems Inc MPA100
OptiMelt Automated Melting Point System, Accurate measurements of melting point and melting range, Automatic and visual determination, Digital movie of each melt, PID-controlled Temperature ramping, Printer output