Learn More
Cole-Parmer MP-250D-PR Programmable Digital Melting Point Apparatus, Beginning/Ending Recording with IQ/OQ Documentation; 100-240 VAC
Supplier:  Cole-Parmer 0301064
Encompass_Preferred
Programmable digital melting point apparatus with beginning/ending recording for pharmacopeia requirements offers selectable ramp rates between 0.2C/minute to 10C/minute with a degree setting of 1C/minute. Store up to two sets (beginning and ending temperatures) of three temperatures in temporary memory; batch memory permanently stores up to 500 sets of results. Unit also connects to computer or printer via RS-232 interface.
Catalog No. 50-196-0378