Learn More
Hach Company MEL/MPN Total Coliform and E. coli Laboratory
Supplier: Hach Company 2569800
This MEL provides all the hardware to carryout the Most Probable Number (MPN) method. In additon to potable water, the MPN method is ideal for wastewater samples and nonpotable samples, because the analyst can accomodate highly turbid samples which would otherwise require filtration. MEL/MPN Total Coliform and E. Coli Laboratory. Includes: portable Incubator, portable UV lamp, marker, thermometer and sampling/filtration consumables for 50 tests. Media is not included (order separately - see manual on Downloads page). For wastewater applications, order dilution water 1430598.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.