Learn More
PIP™ Mega T-Fit™ Corded Ear Plug
T-shape, disposable foam ear plug made of soft polyurethane.
Supplier: PIP™ 267HPF510C
Description
- Innovative 'T-Shape' makes removal simple
- Consistent cellular structure for optimal expansion recovery time for a consistent, uniform fit
- Smooth closed cell outer layer that prevents debris build-up
- Soft and comfortable PowerSoft™ foam supports all day comfort
- Black PVC cord for convenience and to prevent loss of ear plugs
- NRR 32 protection
Specifications
Ear Plugs | |
Class AL | |
No | |
32 NRR | |
T-Shape | |
No | |
EN 352-1 |
Yes | |
Universal | |
1000 Plugs/Cs. | |
Corded | |
Single-use | |
Yes |