Learn More
Medline™ Suture, 0 27 Chromic Gut, CT-1
Supplier:  Medline™ ETH812H
Catalog No. 23-666-009
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Specifications
Specifications
Chromic Monofilament
27 in.
36 Pk.
Yes
CT-1
Taper Point Surgical Suture
70 cm
General soft tissue approximation and ligation, ophthalmic procedures
1/2 Circle
36 mm