Learn More
Electron Microscopy Sciences Medium Heat Cautery Set
Supplier: Electron Microscopy Sciences 7297621
The cautery comes with fully changeable tungsten carbide tips and batteries. Dual power: 1 AA battery for lower power and 2 AA batteries for higher power. Fully autoclavable. Stainless steel construction. Generates 500 to 520 deg C, set includes cautery handle and tips.