Learn More
GEMCO MEDICAL MediSense Glucose and Ketone Control Solution, 4.0 mL vial
Supplier: GEMCO MEDICAL DOPNR
Compatible with FreeStyle Precision Neo, Precision Xtra and Optium EZ meters and the FreeStyle Libre Flash Glucose Monitoring System4.0 mL vial