Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ SafeBasics™ Bouffant Caps

Catalog Number 19910549
Click to view available options
Size:
21 in.
24 in.
Color:
Blue
White
This item is not returnable. View return policy
This item is not returnable. View return policy

Spunbonded polypropylene bouffant caps are worn by healthcare personnel during various procedures to protect patients and materials from transfer of microorganisms and particulates.

  • Fluid repellent material for optimal protection
  • 100 Caps/Box, 5 Boxes/Case
  • Latex Free
Product Type Bouffant
Size 24 in.
Material Spunbond Polypropylene
Disposable Single-use
Latex Free Yes
Model 8003
Sterility Non-sterile
Quantity 100/Pk.
Diameter (English) 24 in.
For Use With (Application) Healthcare
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit