Learn More
Medicus Health Lockable Refrigerator Mount Medication Box with Key Lock, Red
SDP
Supplier:  Medicus Health 2109M3RED
Encompass
∙ Lockable Refrigerator Mount Medication Box with Key Lock securely stores medication that needs to be refrigerated ∙ Rust proof stainless steel frame with pull out drawer keeps items secure. Mounting kit is only accessible when the drawer is open ∙ Medication Lock Boxes with pull-out drawer helps comply with Joint Commission regulations to keep high risk medications locked up at all times ∙ High density polyethylene box resists most solvents & chemicals ∙ All boxes complete with predrilled holes and mounting kits for wire and glass shelves . 10.9"W x 16.5"D x 6"H ∙ Choose from (3) different sizes.
Catalog No. NC2056699