Learn More
Medical Packaging Swab-Pak™ Swabs
Polyester-tipped, general-use swabs
$184.00 - $266.00
Specifications
Tip Material | Dacron |
---|---|
For Use With (Application) | General-use swabs |
Includes | Plastic Tamper-evident labeled transport tube |
Sterility | Sterile |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
22-281659
|
Medical Packaging
SP2D |
Dual Swab with Plastic Shaft |
Pack of 100 for $184.00
Case of 5 PK for $900.00
|
|
|||||
22-281661
|
Medical Packaging
SP7D |
Single Swab with Aluminum Shaft |
Pack of 100 for $266.00
Case of 5 PK for $1,321.00
|
|
|||||
Description
Swabs are contained in a sterile, plastic transport tube with a tamper-evident label
Single swabs have a straight wire shaft on a plastic handle
- Dual swabs have a plastic shaft
Specifications
Dacron | |
General-use swabs | |
Plastic Tamper-evident labeled transport tube | |
Sterile |