Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Medegen Biohazardous Waste Bags, LLDPE Film, Coreless Roll
Meet ASTM™ Dart/Elmendorf and DOT CFR 49 173-197 requirements.
Supplier: Medegen R117
Description
- Red bag with black printing
- Biohazardous/Infectious Waste label with Biohazard symbol printed on bag
Specifications
Biohazardous Waste Bag | |
113.6 L | |
Red, Black | |
LLDPE | |
1.50 mil | |
30.5 in. | |
41 in. |
30 gal. | |
ASTM Dart | |
Biohazardous or infectious waste with biohazard symbol | |
0.038 mm | |
30.5 x 41 in. (77.47 x 104.14cm) | |
77.47 cm | |
104.14 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction