Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Medique Medi-Seltzer™ Tablets
Effervescent pain reliever and antacid with specially buffered aspirin.
$16.00 - $29.75
Specifications
Product Type | Buffered Aspirin Tablets |
---|---|
Active Ingredients | Aspirin 324 mg, Heat Treated Sodium Bicarbonate 1916 mg, Citric Acid (in water) 1000 mg |
Format | Tablet |
For Use With (Application) | Antacid and pain reliever |
Recommended Age | 12 years and older |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
19-090-789
|
Medique
13520 |
2/Pk.,18/Box |
Each for $16.00
Case of 36 EA for $512.00 |
|
19-090-788
|
Medique
13524 |
2/Pk., 36/Box |
Each for $29.75
Case of 36 EA for $971.00 |
|
Description
- Antacid and pain reliever
- Each Tablet contains Aspirin 324mg; Heat Treated Sodium Bicarbonate 1,916mg; Citric Acid (in water) 1,000mg
Specifications
Buffered Aspirin Tablets | |
Tablet | |
12 years and older |
Aspirin 324 mg, Heat Treated Sodium Bicarbonate 1916 mg, Citric Acid (in water) 1000 mg | |
Antacid and pain reliever | |
59°F to 86°F (15°C to 30°C) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title