Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meclizine Hydrochloride, USP, 97-102%, Spectrum™ Chemical
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
Description
Spectrum™ Chemical Meclizine Hydrochloride, USP
Specifications
CAS Max % | 100% |
Ignition Residue | 0.1% |
Packaging | Amber Glass Bottle |
Quantity | 1 kg |
Percent Purity | 97 to 102% |
Grade | USP |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction