Learn More
Seca Corp 216 Mechanical wall-mounted measuring rod for children and adults, IN/CM
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Seca Corp 2161814009
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The seca 216 stadiometer has a variable scale with a range of 1.4 to 90 inches / 3.5 - 230 cm. The plastic frame is easily mounted on the wall at the desired height. As the user can choose the measurement range, to adapt to a patient population mainly of children or adults (or both). The results are clearly displayed in a window on the side of the rod and held in place by the locking screw on the head piece. Units of measure are printed in centimeters on one side of the tape and centimeters and inches on the other. • Easy-to-read measurement scale • Adjustable head positioner • Choice of two units of measurement • Simple assembly • Measuring range: max. 55.3" / 138 cm, variable from 1.4 – 90" / 3.5 – 230 cm • Graduation: 1/16 inch / 1 mm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.