Learn More
Electron Microscopy Sciences EMS Model IAM-8, Multi Grids Standard
Supplier: Electron Microscopy Sciences 602168PG
A uniqulye designed standard with 4 different grid size patterns: 0.5 x 0.5 mm grid with 25 um pitch; 1.0 x 1.0 mm grid with 50 um pitch; 2.0 x 2.0 grid with 100 um pith; and 4.0 x 4.0 mm grid with 200 um pitch. Also includes a Star Target with 15 degree angle increments. This standard can be used to test not only the overall frame distortion issue, but also linear distances for a wide variety of magnification ranges from 1x to 1000x power.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.