Learn More
Morwell MD Biosciences MDB ELISA ACCESSORY KIT
Supplier: Morwell MD Biosciences M036080
MonELISA Ancillary Reagent Kit containing all buffers / diluents / stop solution for the performance of MonELISA assay series. 5 Plates