Learn More
MCR Safety Storm™Safety Glasses
Click to view available options
Lens Tint:
Green 3.0
Green 5.0
Description
Wraparound lens constructed of lightweight, scratch resistant polycarbonate, that filters 99.9% of U.V. rays, delivers advanced visual acuity and protection.
- Available in a variety of frame colors and lens combinations
- Single wraparound dual focal point lens with integral side shields
- 5 position ratchet action bayonet temples
- Exclusive Duramass™ scratch-resistant coating
- Non-slip, soft temple material
Specifications
Lens Tint | Green 5.0 |
Product Type | Safety Glasses |
Frame Color | Black |
Frame Material | Nylon |