Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCR Safety NitriMed™ -XTRA Nitrile Gloves
Click to view available options
Small
Description
- 12 inch length and 6 mil thickness
- Medical Grade, Class 1
- Powder free
- Rolled cuff
- Latex-free
- Resistant to punctures and tears
- Textured grip
Specifications
Certifications/Compliance | Rated as Class 1 Medical Device |
Certifications/Compliance (Europe) | Class B according the EN374:2016 standard |
Chemotherapeutic Rated | No |
Applications | Healthcare, Laboratory |
Color | Blue |
Disposable | Yes |
Exterior Finish | Partially Textured |
Laboratory/Cleanroom Use | No |
Latex Free | Yes |
Material | Nitrile |
Show More |