Learn More
Anton Paar Modular Compact Polarimeter – MCP 150
Supplier: Anton Paar 160796
The MCP 150 polarimeter provides proven technology packed into a compact device which fits into any laboratory. The instrument is extremely easy to operate, provides maximum comfort, high security and full compliance with all national and international standards such as 21 CFR Part 11. The MCP 150 is the right choice for sophisticated analysis in the pharmaceutical, cosmetics, food and chemical industries as well as R&D and medical applications.- 21 CFR Part 11 compliance with electronic signature and user group administration- Built-in Peltier temperature control- Wireless automatic identification of sample cells and quartz control plates via RFIDMeasuring scales: °Optical Rotation, °Specific Rotation, %Concentration, user-defined Measuring range °Optical RotationRequires a Measurement Cell: NC1779947 or NC1710887 or NC1818318 or NC1818320Accessories: NC1779946, NC1818321, NC1818322
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.