Learn More
Anton Paar Modular Compact Polarimeter – MCP 100
Supplier:  Anton Paar 143911
Encompass_Preferred
The MCP 100 polarimeter provides proven technology packed into a compact polarimeter which fits into any laboratory. The instrument is extremely easy to operate and also provides maximum comfort and high security. The combination of quality, all important features and modern data management makes MCP 100 the most efficient high precision compact polarimeter on the market. - MCP 100 covers the application requirements of the pharmaceutical, cosmetics, food and chemical industries as well as R&D and medical applications.- Built-in Peltier temperature control- Wireless automatic identification of sample cells and quartz control plates via RFIDMeasuring scales: °Optical Rotation, °Specific Rotation, %ConcentrationMeasuring range °Optical Rotation- Range ±89.9°- Resolution 0.001°- Accuracy ±0.01°- Repeatability ±0.01°Temperature- Range 20°C and 25°C- Accuracy ±0.2 °CRequires a Measurement Cell: NC1818309 or NC1818310 or NC1818312 or NC1818313Accessories: NC1818315, NC1818317
Catalog No. NC1818306