Learn More
Key Scientific Products McFarland Latex Standards Tube, 0.5 12MLMCF
Supplier: Key Scientific Products HML05
Prepared from suspensions of uniform latex particles with similar absorbance as the original barium sulfate standards. Longer shelf-life, better uniformity, and less clumping than the original standards due to reduced light sensitivity.