Learn More
Advantec MFS, Inc Sterile MCE Membrane Filters, Advantec MFS, 47 mm, White, 0.45 um, Gridded, Individual, 100/pk
Supplier: Advantec MFS, Inc A045H047A
Mixed Cellulose Esters (MCE) membrane is a general purpose filter used in many applications for the microbiological examination of water, wastewater, pharmaceuticals, food, and beverages and air monitoring. MCE membranes offer high recovery rates and superior flow rates.