Learn More
Advantec MFS, Inc Mixed Cellulose Esters(MCE) Membrane Filter 47mm, 1000/pk
Supplier: Advantec MFS, Inc A045H047W
Mixed Cellulose Esters(MCE) Membrane Filter. Industry standard for applications in microbiology, sterilizing biological fluids, air monitoring. High protein binding and flow rates. Triton-free. 0.45um; Grid; 47mm; Sterile indiv. wrapped.