Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mopec MB150 Grossing Station Countertop Dry Unit
Standard features bring production to life and optional accessories customize your grossing station to your unique requirements. Mopec™ MB150 Grossing Station Contertop Dry Unit uses standard features and optional accessories to make this workstation ideal for any work environment.
Supplier: Mopec MB150
Description
- Recessed stainless steel work surface
Removable polyethylene cutting board 58.4 x 40.6 x 2cm - Washable ruler with inch/centimeter markings
- Ruler is located on the inside lip of the workstation
- Magnetic tool bar holds instruments when not in use
- One large and one small stainless steel organizer bin
- 4 leveling pads
- Stainless steel c-fold paper towel dispenser
- Waterproof light with 2 LED bulbs
- Back draft ventilation is designed to safely and efficiently remove formaldehyde fumes and hazardous chemical vapors
- Equipped with a factory installed exhaust duct which can be connected to any in-house ventilation system
- Electrical work accomplished at factory
- Only trade hook-up required
Warranty and Services
1 Year Warranty
Specifications
Grossing Station | |
60 Hz | |
Back Draft Ventilation | |
116 cm |
GFCI Electrical Receptacle | |
115V | |
122cm | |
74cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction