Learn More
BD MAX™ Molecular Urine Transport Kit
BD Molecular Urine Transport Kit is intended to be used in clinical settings for collection, preservation and transport of urine specimens.
Supplier: BD 443924
Description
- BD Molecular Urine Transport Kit is intended to be used in clinical settings for collection, preservation and transport of urine specimens
- For use with BD MAX™ System
- Tubes containing 0.5 mL Sample Buffer
Specifications
Urine Preservative Tube | |
Snap Cap | |
No | |
Non-sterile |
Yellow | |
Single-use | |
Polystyrene | |
Yes |