Learn More
ProteinSimple MAURICE CIEF ELECTROLYTE TANK
Supplier: ProteinSimple 046123
Electrolyte tank caps, 20 mm. Red is used for Anolyte tank and the Grey cap is used for the Catholyte tank. 5 pairs/pk