Learn More
Corning™ Matrigel™ hESC-Qualified Matrix, LDEV-Free
Solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
Supplier: Corning™ 356278
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
Specifications
Store at -20°C. Avoid multiple freeze-thaws. | |
No LDEV | |
50 mL (5 mL x 10) |
hESC-qualified Matrix | |
Cell Growth and Differentiation, Metabolism/Toxicology Studies, Invasion Assays, In Vitro and In Vivo Angiogenesis Assays, In Vivo Angiogenesis Studies and Augmentation of Tumors in immunosuppressed Mice | |
Engelbreth-Holm-Swarm Mouse Tumor |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.