Learn More
Buehler MASTERTEX PSA 12IN 10/PK
Supplier:  Buehler 407742
Encompass
This item has a minimum qty of 4 per supplier requirements.
MasterTex is a soft, low nap, synthetic velvet cloth. It is used for final oxide polishing particularly for very soft materials. Typical applications include but are not limited to soft non-ferrous materials and microelectronics packages.
Catalog No. NC1718152