Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Variable-Speed Digital Drive with Remote I/O, 1 to 100 rpm; 90 to 260 VAC
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX0752830
These accurate analog console drives have powerful motors, speed display, and ¿0.25% speed control for precise flow regulation¿ideal for metering applications. Remote speed control requires DB9 male connector 07595-45 or cable assembly 07595-47. Use remote connector for control of start/stop via contact closure or open collector (5 V TTL); control start/stop via footswitch 77595-35 or handheld remote 07528-80.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.