Learn More
Accuflow Systems Masterflex L/S Digital Drive, 600 RPM, 115/230 VAC 0
Supplier: Accuflow Systems 0752220
INCLUDES: 6-ft (1.8-m) line cord with IEC 320/CEE22 socket. These models are shipped with country specific plug/cord set; Pump Only. Precision digital drive offers a maintenance-free high-accuracy motor and a graphical interface – makes setup and operation easier than ever! Flow rate 0.001 to 3400 mL/min with L/S tubing. Flow rate depends on drive rpm and tubing size. Graphical LCD shows four operating modes continuous run, timed dispense, copy dispense, volume dispense. View pump performance continually.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.