Learn More
Epredia™ Mask Cytoslides
Used for immunofluorescence. Masking blocks out UV light to enhance microscopic viewing.
$7.81
Catalog Number | Mfr. No. | Color | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Description | Price | Quantity & Availability | ||||
59-910-57
|
Epredia™
5991057 |
Black | Single circle |
Pack of 100 for $132.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
B5991050
|
Epredia™
B5991050 |
Blue | Double circle |
Pack of 100
for $7.81
Save 95% Reg : |
|
||||
Description
- For immunofluorescence
- Blocks out UV light
Specifications
100 |