Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ MAS™ Urinalysis Controls
Combine dipstick methods and microscopic components with bi-level, liquid assayed MAS Urinalysis Controls, assayed for most reagent strips and strip systems.
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Description | Urinalysis Control, Assayed |
Detectable Analytes | Bilirubin, Blood, Creatinine, Crystals, Glucose, hCG, Ketones, Leukocyte Esterase, Microalbumin, Nitrite, Osmolality, pH, Potassium, Protein, Red Blood Cells, Sodium, Specific Gravity, Urobilinogen, White Blood Cells |
Sample Type | Urine |
Type | Control |
Catalog Number | Mfr. No. | Level | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Level | Quantity | Price | Quantity & Availability | ||||
23-030125
|
Thermo Scientific™
024222 |
1 | 15 mL | N/A | N/A | ||||
This product is regulated by the federal government, and may not be ordered online.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-030128
|
Thermo Scientific™
024225 |
1 | 60 mL | N/A | N/A | ||||
This product is regulated by the federal government, and may not be ordered online.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-030127
|
Thermo Scientific™
024224 |
3 | 15 mL | N/A | N/A | ||||
This product is regulated by the federal government, and may not be ordered online.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-030131
|
Thermo Scientific™
024227 |
3 | 60 mL | N/A | N/A | ||||
This product is regulated by the federal government, and may not be ordered online.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
Description
Combine dipstick methods and microscopic components with bi-level, liquid assayed Thermo Scientific™ MAS™ Urinalysis Controls. This control is assayed for most major reagent strips and strip systems.
- Human urine matrix
Specifications
2°C to 8°C | |
Bilirubin, Blood, Creatinine, Crystals, Glucose, hCG, Ketones, Leukocyte Esterase, Microalbumin, Nitrite, Osmolality, pH, Potassium, Protein, Red Blood Cells, Sodium, Specific Gravity, Urobilinogen, White Blood Cells | |
Control |
Urinalysis Control, Assayed | |
Urine | |
4 Pk. |
Safety and Handling
ShelfLife : 12 Months
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title