Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ MAS™ Diabetes Control
Monitor hemoglobin A1c methods with Thermo Scientific™ MAS™ Diabetes Control, a bi-level control available in a convenient Vista™ vial format.
Specifications
Content And Storage | Two year shelf life at -25° to -15°C; 30 days open vial at 2° to 8°C |
---|---|
Detectable Analytes | Hemoglobin A1c |
Sample Type | Whole Blood |
Type | Control |
Level | Bi-level |
Catalog Number | Mfr. No. | Description | Level | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Level | Quantity | Price | Quantity & Availability | |||
23-640-25
|
Thermo Scientific™
DBCLMP |
Multi-pack | Bi-level | 6 x 1 mL |
Pack of 6 for $732.50
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-640-27
|
Thermo Scientific™
DBCLV |
Vista Vials | Bi-level | 6 x 2 mL | N/A | N/A | |||
This product is regulated by the federal government, and may not be ordered online.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
Description
Monitor hemoglobin A1c methods with Thermo Scientific™ MAS™ Diabetes Control, a bi-level control available in a convenient Vista™ vial format. This read-to-use, liquid assayed diabetes control has been assayed for most major instrument systems.
- Human whole blood matrix
- Assayed values available for most major analyzers
Specifications
Two year shelf life at -25° to -15°C; 30 days open vial at 2° to 8°C | |
Whole Blood | |
Bi-level |
Hemoglobin A1c | |
Control | |
6 Pk. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title