Learn More
Advanced Materials Technology MARVELXACT PEEKSIL 25 ID X
Supplier: Advanced Materials Technology PS7025600
Advanced Materials Technology’s MarvelXACT finger-tight UHPLC connectors with volume definitions easily identifies dispersion volume contribution. Haptic “click” indicates optimum torque and zero dead volume connection. Connector and tubing for both pre and post column installation