Learn More
Gawet Marble and Granite Vermont Marble™ Slab, 18″ x 14″ x 2″ thick
Supplier: Gawet Marble and Granite 18X14X2
The Vermont Marble™ Slab is for use on laboratory furniture. It substantially reduces vibration and shock where a Vermont Marble™ Balance Table cannot be used.– Standard Matte Machined Finish– Includes set of 4 anti-vibration damping pads