Learn More
Diversified Spaces™ Maple Table with Plain Apron and Maple Top: Leg Brace and Stretcher Option
Constructed from select northern maple, maximum length laminated strips, and finished with sealer and multiple coats of lacquer. Diversified Spaces™ Maple Tables with Maple Tops are used in four-station workbenches, wall benches, and heavy-duty workbenc
$991.00 - $1668.00
Specifications
Adjustable | No |
---|---|
Certifications/Compliance | SEFA compliant and MAS Certified Green™ |
Finish | Chemical resistant, earth-friendly UV |
Includes | Leg Brace and Stretcher |
Load Bearing Capacity (English) | 500 lb. |
Catalog Number | Mfr. No. | Dimensions (L x W) | Height (English) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Height (English) | Includes | Price | Quantity & Availability | |||
S42139
|
Diversified Spaces™
P7605M34S |
24 x 60 in. (60.96 x 152.4 cm) | 34 in. | Leg Brace and Stretcher |
Each for $1,221.00
|
|
|||
S42132
|
Diversified Spaces™
P7605M30S |
24 x 60 in. (60.96 x 152.4 cm) | 30 in. | Leg Brace and Stretcher |
Each for $1,146.00
|
|
|||
S41939
|
Diversified Spaces™
P7305M36S |
24 x 72 in. (60.96 x 182.88 cm) | 36 in. | Leg Brace and Stretcher |
Each for $1,382.00
|
|
|||
S42695
|
Diversified Spaces™
P7105M30S |
24 x 48 in. (60.96 x 121.92 cm) | 30 in. | Leg Brace and Stretcher |
Each for $991.00
|
|
|||
S47839
|
Diversified Spaces™
P7145M34S |
30 x 60 in. (76.2 x 152.4 cm) | 34 in. | Leg Brace and Stretcher |
Each for $1,259.00
|
|
|||
S47479
|
Diversified Spaces™
P7105M34S |
24 x 48 in. (60.96 x 121.92 cm) | 34 in. | Leg Brace and Stretcher |
Each for $1,066.00
|
|
|||
S47832
|
Diversified Spaces™
P7145M30S |
30 x 60 in. (76.2 x 152.4 cm) | 30 in. | Leg Brace and Stretcher |
Each for $1,185.00
|
|
|||
S47484
|
Diversified Spaces™
P7105M36S |
24 x 48 in. (60.96 x 121.92 cm) | 36 in. | Leg Brace and Stretcher |
Each for $1,066.00
|
|
|||
S47846
|
Diversified Spaces™
P7145M36S |
30 x 60 in. (76.2 x 152.4 cm) | 36 in. | Leg Brace and Stretcher |
Each for $1,259.00
|
|
|||
S47981
|
Diversified Spaces™
P7155M30S |
30 x 72 in. | 30 in. | Leg Brace and Stretcher |
Each for $1,594.00
|
|
|||
S47987
|
Diversified Spaces™
P7155M34S |
30 x 72 in. | 34 in. | Leg Brace and Stretcher |
Each for $1,668.00
|
|
|||
S48454
|
Diversified Spaces™
P7205M30S |
24 x 54 in. (60.96 x 137.16 cm) | 30 in. | Leg Brace and Stretcher |
Each for $1,067.00
|
|
|||
S47989
|
Diversified Spaces™
P7155M36S |
30 x 72 in. | 36 in. | Leg Brace and Stretcher |
Each for $1,668.00
|
|
|||
S48461
|
Diversified Spaces™
P7205M34S |
24 x 54 in. (60.96 x 137.16 cm) | 34 in. | Leg Brace and Stretcher |
Each for $1,142.00
|
|
|||
S48468
|
Diversified Spaces™
P7205M36S |
24 x 54 in. (60.96 x 137.16 cm) | 36 in. | Leg Brace and Stretcher |
Each for $1,142.00
|
|
|||
Description
- 1.75 in. Maple Table Surface
- 2.25 in. solid square legs, with 5/16 in.hanger bolts installed, supplied nuts and washers
- Aprons are 13/16 in.D x 4.5 in.H with heavy-duty 13 gauge steel corner braces
- Diversified Spaces™ is SEFA compliant and MAS Certified Green
- Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
- Includes leg braces and stretcher
- Limited Lifetime Warranty
- Made in the USA
- Minimal assembly required; legs need to be attached
- Solid Maple Construction
Specifications
No | |
Chemical resistant, earth-friendly UV | |
500 lb. | |
Plain Apron | |
Plain Apron Table |
SEFA compliant and MAS Certified Green™ | |
Leg Brace and Stretcher | |
226.79 kg | |
Maple | |
Maple |