Learn More
Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
Supplier: Thermo Scientific™ CR8C
Description
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.
Manual CrimpersLightweight, precision crimpers attach seals to chromatography or serum vials. Available for standard aluminum or flip-off seals.
- Factory set to properly crimp standard-thickness seals
- Adjustable handle setting screw allows proper crimps with septa of varying thickness
Manual Decrimpers
Forged pliers feature a fitted gripping surface designed to allow removal of the listed sizes of seals.
- Simply grip pliers around top of seal, squeeze and pry upward to remove
Manual Decrimping Pliers
Place decrimping head squarely on seal and squeeze.
- Minimizes damage to vial rim with chromatography or serum vials
- Removes standard aluminum seals and many steel seals
Specifications
8mm Standard Seal, National Vial, Chromacol Vial | |
Industrial Chromatography | |
Manual Crimper | |
1 Ea. |
8 mm | |
Chromacol | |
Manual Crimper, Standard |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.