Learn More
Chemglass Life Sciences Glas-Col Mantle Extension Support for 50mL Series O Mantle, 3 3/4in ID
SDP
Supplier:  Chemglass Life Sciences CG1001001
Encompass_Preferred
Glas-Col Mantle Extension Support for 50mL Series O Mantle, 3 3/4in ID. This lattice mount support holds spherical and hemispherical Glas-Col Series O mantles up to 5-liter capacity. The steel support, with its fabric strap basket, is strong, handy and durable. Attaches easily to stable ring stands or distillation racks. Supports are painted gray. This mantle support is made from 3/8in diameter rod.
Catalog No. CG1001001
Due to product restrictions, please to purchase or view availability for this product.