Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Manganese Chloride, 1.0M Solution (Clear, Lt.-Pink Solution/Molecular Biology), Fisher BioReagents
Click to view available options
Poly Bottle; 100mL
Description
Manganese Chloride is used for dC and dG tailing of DNA.
Specifications
Color | Red |
CAS | 7773-01-5 |
Molecular Formula | Cl2Mn |
MDL Number | MFCD00011114 |
Concentration | 0.99 to 1.01M |
Merck Index | 15, 5793 |
Synonym | manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline |
InChI Key | GLFNIEUTAYBVOC-UHFFFAOYSA-L |
SMILES | [Cl-].[Cl-].[Mn++] |
IUPAC Name | manganese(2+) dichloride |
Show More |
Safety and Handling
Recommended Storage | RT |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction