Learn More
Fine Science Tools MANDREL FOR MICRO DRILL
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Fine Science Tools 1800018
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Product: Mandrel for Micro Drill/ Shaft Diameter: 2.3mm/ Alloy/Material: Stainless Steel/ Length: 45mm