Learn More
Avantor™ Mallinckrodt Baker, CHEM SOLV glass cleaner
Supplier: Avantor™ 215704
Specifications
Poly bottle | |
Cleaning applications where Chromic Acid would normally be used, effective on Silicone greases, dried Blood and other organic residues | |
6 qt. |