Learn More
MilliporeSigma™ Supelco™ Magnetic Stir Bar
Magnetic stir bar may be used for stirring samples in the stir-bar sorptive extraction (SBSE) of volatile components of malt whisky, analyzed using gas chromatography coupled with mass spectrometry (GC-MS)
$186.40
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
11-102-1610
|
MilliporeSigma™ Supelco™
23226 |
0.1 to 2 mL Vessels |
Each for $186.40
|
|
|||||
11-102-1611
|
MilliporeSigma™ Supelco™
23227 |
3 to 5 mL Vessels |
Each for $186.40
|
|
|||||
Description
Specifications
PTFE | |
6/Pk. |