Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium sulfate, 1M aq. soln., sterile-filtered, Thermo Scientific Chemicals

Catalog Number AAJ61030EQE
Click to view available options
Quantity:
125 mL
250 mL
7487-88-9
MgO4S
120.36
MFCD00011110
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
24083
CHEBI:32599
magnesium;sulfate
[Mg++].[O-]S([O-])(=O)=O
7487-88-9
MgO4S
120.36
MFCD00011110
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
24083
CHEBI:32599
magnesium;sulfate
[Mg++].[O-]S([O-])(=O)=O

Magnesium sulfate is applied in replacement therapy for hypomagnesemia. It can be used orally as a laxative to relieve occasional constipation, and to treat low levels of magnesium. It works by replacing magnesium in the body and increasing water in the intestines. It is the first-line antiarrhythmic agent for torsades de pointes in cardiac arrest under the 2005 ECC guidelines and for managing quinidine-induced arrhythmias. It is effective in decreasing the risk that pre-eclampsia progresses to eclampsia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Fully miscible in water. Soluble in alcohol, ether (11.6 mg/ml at 18°C) and glycerin. Insoluble in acetone.

Notes
Stable under recommended storage conditions. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Physical Form Liquid
Quantity 125 mL
Linear Formula MgSO4
Solubility Information Fully miscible in water. Soluble in alcohol,ether (11.6mg/ml at 18°C) and glycerin. Insoluble in acetone.
Formula Weight 120.37
Concentration or Composition (by Analyte or Components) 1M aq. soln.
Chemical Name or Material Magnesium sulfate, sterile
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit