Learn More
Bioworld Magenta GA-7 Plant Culture Box w/ Lid 100 units
Supplier:  Bioworld 309300072
Encompass_Preferred
Made of polycarbonate, non-sterile vessel with polypropylene lid. Magenta boxes are vessels used in the plant culture process. The boxes are autoclavable and reusable. They are sturdy and leak free. The clear surface allows one to view samples without disrupting growth. The boxes have a growing area of 6.125 sq. inches. Dimensions: 3 x 3 x 4". 100 units/cs
Catalog No. 50-255-176